Anti-NGFRAP1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306179EA 600 EUR
ORIGTA306179
Anti-NGFRAP1 Rabbit Polyclonal Antibody
Antikörper
  • Recommended Dilutions: Western Blot: 0.5-2 μg/ml

Type: Primary
Antigen: NGFRAP1 (nerve growth factor receptor (TNFRSF16) associated protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR