Anti-IL1RL1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306180EA 600 EUR
ORIGTA306180
Anti-IL1RL1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: IL1RL1 (interleukin 1 receptor-like 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR