Anti-IRF3 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306191EA 595 EUR
ORIGTA306191
Anti-IRF3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: IRF3 (interferon regulatory factor 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR