Drucken

Anti-CASP6 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306203EA 482 EUR InStock
ORIGTA306203
Anti-CASP6 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1 and 2 ?g/ml; Immunochemistry: 1 ?g/ml; Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: CASP6 (caspase 6, apoptosis-related cysteine peptidase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more