Anti-C1QTNF7 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306240EA 600 EUR
ORIGTA306240
Anti-C1QTNF7 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5 to 2 ?g/ml; Immunochemistry: 0.5 ?g/ml; Immunofluorescence: 2 ?g/ml

Type: Primary
Antigen: C1QTNF7 (C1q and tumor necrosis factor related protein 7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR