Anti-LAMP2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306261EA 600 EUR
ORIGTA306261
Anti-LAMP2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunochemistry: 10?g/ml

Type: Primary
Antigen: LAMP2 (lysosomal-associated membrane protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR