Anti-GOPC Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306263EA 600 EUR
ORIGTA306263
Anti-GOPC Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: GOPC (golgi-associated PDZ and coiled-coil motif containing)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR