Anti-TLR6 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306272EA 595 EUR
ORIGTA306272
Anti-TLR6 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-2 ?g/ml; Immunochemistry: 2 ?g/ml

Type: Primary
Antigen: TLR6 (toll-like receptor 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR