Anti-ATF6 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306287EA 600 EUR
ORIGTA306287
Anti-ATF6 Rabbit Polyclonal Antibody
Antikörper
  • Recommended Dilutions: Western Blot: 0.5-1 μg/ml; Immunochemistry: 10 μg/ml

Type: Primary
Antigen: ATF6 (activating transcription factor 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR