Anti-TLR9 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306291EA 494 EUR
ORIGTA306291
Anti-TLR9 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-2 ?g/ml; Immunochemistry: 2 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: TLR9 (toll-like receptor 9)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR