Drucken

Anti-ACTB Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306308EA 482 EUR InStock
ORIGTA306308
Anti-ACTB Rabbit Polyclonal Antibody
Antikörper
  • Recommended Dilutions: Western Blot: 0.5-2 μg/ml; Immunochemistry: 10 μg/ml; Immunofluorescence: 20 μg/ml

Type: Primary
Antigen: ACTB (actin, beta)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more