Anti-IL27 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306317EA 600 EUR
ORIGTA306317
Anti-IL27 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 2-4 ?g/ml

Type: Primary
Antigen: IL27 (interleukin 27)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR