Drucken

Anti-BAG1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306337EA 482 EUR InStock
ORIGTA306337
Anti-BAG1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: BAG1 (BCL2-associated athanogene)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more