Anti-SCO2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306396EA 600 EUR
ORIGTA306396
Anti-SCO2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-1?g/ml

Type: Primary
Antigen: SCO2 (SCO2 cytochrome c oxidase assembly protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR