Anti-DARC Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306405EA 600 EUR
ORIGTA306405
Anti-DARC Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: DARC (Duffy blood group, atypical chemokine receptor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR