Anti-HUWE1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306429EA 600 EUR
ORIGTA306429
Anti-HUWE1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunochemistry: 5 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: HUWE1 (HECT, UBA and WWE domain containing 1, E3 ubiquitin protein ligase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR