Anti-VIPR2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA308447EA 642 EUR
ORIGTA308447
Anti-VIPR2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot:1:500-1:3000

Type: Primary
Antigen: VIPR2 (vasoactive intestinal peptide receptor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR