Anti-SHANK2 Mouse Monoclonal Antibody [clone: S23b-6]

Lieferant: OriGene
ORIGTA309475EA 678 EUR
ORIGTA309475
Anti-SHANK2 Mouse Monoclonal Antibody [clone: S23b-6]
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: SHANK2 (SH3 and multiple ankyrin repeat domains 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR