Anti-EMCN Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA312706EA 564 EUR
ORIGTA312706
Anti-EMCN Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:1000

Type: Primary
Antigen: EMCN (endomucin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR