Anti-ZHX2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA312720EA 564 EUR
ORIGTA312720
Anti-ZHX2 Rabbit Polyclonal Antibody
Antikörper
  • Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:10000

Type: Primary
Antigen: ZHX2 (zinc fingers and homeoboxes 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR