Anti-BCAR1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA312725EA 564 EUR
ORIGTA312725
Anti-BCAR1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-3000; Immunochemistry: 1:50-1:100; ELISA: 1:5000

Type: Primary
Antigen: BCAR1 (breast cancer anti-estrogen resistance 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR