Anti-RABL2A Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA312776EA 552 EUR
ORIGTA312776
Anti-RABL2A Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: RABL2A (RAB, member of RAS oncogene family-like 2A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR