Anti-TSC22D1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA312827EA 552 EUR
ORIGTA312827
Anti-TSC22D1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: TSC22D1 (TSC22 domain family, member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR