Anti-OR52E5 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA312845EA 532 EUR
ORIGTA312845
Anti-OR52E5 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: OR52E5 (olfactory receptor, family 52, subfamily E, member 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR