Anti-STAT2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA312889EA 564 EUR
ORIGTA312889
Anti-STAT2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:50-100; ELISA: 1:20000

Type: Primary
Antigen: STAT2 (signal transducer and activator of transcription 2, 113kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR