Anti-JUND Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA312901EA 532 EUR
ORIGTA312901
Anti-JUND Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; ELISA: 1:20000; Immunoprecipitation: Various Dilution

Type: Primary
Antigen: JUND (jun D proto-oncogene)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR