Anti-CDC42BPA Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313011EA 564 EUR
ORIGTA313011
Anti-CDC42BPA Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: CDC42BPA (CDC42 binding protein kinase alpha (DMPK-like))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR