Anti-STAT5A Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313032EA 552 EUR
ORIGTA313032
Anti-STAT5A Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:1000

Type: Primary
Antigen: STAT5A (signal transducer and activator of transcription 5A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR