Anti-MAP3K11 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313098EA 552 EUR
ORIGTA313098
Anti-MAP3K11 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: MAP3K11 (mitogen-activated protein kinase kinase kinase 11)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR