Anti-CDC25B Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313189EA 552 EUR
ORIGTA313189
Anti-CDC25B Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1000; ELISA: 1:20000

Type: Primary
Antigen: CDC25B (cell division cycle 25B)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR