Anti-TOP2A Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313198EA 552 EUR
ORIGTA313198
Anti-TOP2A Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: TOP2A (topoisomerase (DNA) II alpha 170kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR