Anti-PMS2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313327EA 552 EUR
ORIGTA313327
Anti-PMS2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: PMS2 (PMS2 postmeiotic segregation increased 2 (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR