Anti-ABCB7 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313408EA 552 EUR
ORIGTA313408
Anti-ABCB7 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:5000

Type: Primary
Antigen: ABCB7 (ATP-binding cassette, sub-family B (MDR/TAP), member 7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR