Anti-ADORA3 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313433EA 543 EUR
ORIGTA313433
Anti-ADORA3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:5000

Type: Primary
Antigen: ADORA3 (adenosine A3 receptor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR