Anti-APLP2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313464EA 552 EUR
ORIGTA313464
Anti-APLP2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: APLP2 (amyloid beta (A4) precursor-like protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR