Anti-CAMK2G Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313553EA 552 EUR
ORIGTA313553
Anti-CAMK2G Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: CAMK2G (calcium/calmodulin-dependent protein kinase II gamma)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR