Anti-CDH15 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA313602
ORIGTA313602EA 564 EUR
ORIGTA313602
Anti-CDH15 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: CDH15 (cadherin 15, type 1, M-cadherin (myotubule))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR