Anti-COL4A3 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313650EA 564 EUR
ORIGTA313650
Anti-COL4A3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:1000

Type: Primary
Antigen: COL4A3 (collagen, type IV, alpha 3 (Goodpasture antigen))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR