Anti-COL6A3 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313660EA 564 EUR
ORIGTA313660
Anti-COL6A3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:40000

Type: Primary
Antigen: COL6A3 (collagen, type VI, alpha 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR