Anti-CPB2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313671EA 564 EUR
ORIGTA313671
Anti-CPB2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000 References:

Type: Primary
Antigen: CPB2 (carboxypeptidase B2 (plasma))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR