Anti-CSF2RA Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313697EA 552 EUR
ORIGTA313697
Anti-CSF2RA Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:1000; ELISA: 1:20000

Type: Primary
Antigen: CSF2RA (colony stimulating factor 2 receptor, alpha, low-affinity (granulocyte-macrophage))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR