Anti-VCAN Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313704EA 564 EUR
ORIGTA313704
Anti-VCAN Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:10000

Type: Primary
Antigen: VCAN (versican)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR