Anti-CSTA Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313707EA 564 EUR
ORIGTA313707
Anti-CSTA Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:5000

Type: Primary
Antigen: CSTA (cystatin A (stefin A))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR