Anti-CX3CR1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313715EA 552 EUR
ORIGTA313715
Anti-CX3CR1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:5000

Type: Primary
Antigen: CX3CR1 (chemokine (C-X3-C motif) receptor 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR