Anti-CYP2A7 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313719EA 552 EUR
ORIGTA313719
Anti-CYP2A7 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: CYP2A7 (cytochrome P450, family 2, subfamily A, polypeptide 7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR