Anti-DHX8 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313749EA 552 EUR
ORIGTA313749
Anti-DHX8 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:5000

Type: Primary
Antigen: DHX8 (DEAH (Asp-Glu-Ala-His) box polypeptide 8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR