Anti-DNMT3B Rabbit Polyclonal Antibody

Lieferant: OriGene
TA313761
ORIGTA313761EA 564 EUR
ORIGTA313761
Anti-DNMT3B Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: DNMT3B (DNA (cytosine-5-)-methyltransferase 3 beta)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR