Anti-CELSR3 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313791EA 552 EUR
ORIGTA313791
Anti-CELSR3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: CELSR3 (cadherin, EGF LAG seven-pass G-type receptor 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR