Anti-F12 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313860EA 552 EUR
ORIGTA313860
Anti-F12 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: F12 (coagulation factor XII (Hageman factor))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR