Anti-FOXE1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA313878EA 552 EUR
ORIGTA313878
Anti-FOXE1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: FOXE1 (forkhead box E1 (thyroid transcription factor 2))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR